siRNA Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181489.5 | NP_852466.1 | 8 | 2893 | ||
| XM_005265102.2 | XP_005265159.1 | 7 | 2751 | |||
| XM_005265103.3 | XP_005265160.1 | 5 | 2023 | |||
| XM_011533674.2 | XP_011531976.1 | 7 | 2716 | |||
| XM_011533675.2 | XP_011531977.1 | 5 | 1987 | |||
| XM_017006345.1 | XP_016861834.1 | 7 | 2772 | |||
| XM_017006346.1 | XP_016861835.1 | 7 | 2715 | |||
| GenBank mRNA | AY262260.1 | - | 6 | 2696 | ||
| AY295873.2 | - | 8 | 2893 | |||
| BC101486.1 | - | 6 | 2607 | 8068992 | ||
| BC143439.1 | - | 6 | 2572 | 9051947 | ||

ZNF445
Genomic Map