This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF445 SIRNA Assays ›
Gene Symbol:
ZNF445
Gene Name:
zinc finger protein 445
Gene Aliases:
ZKSCAN15, ZNF168, ZSCAN47
Chromosome Location:
Chr.3: 44439770 - 44477670 on Build GRCh38
UniGene ID:
Hs.250481
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s51458

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF445
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181489.5 NP_852466.1 8 1974
XM_005265102.2 XP_005265159.1 7 1832
XM_005265103.3 XP_005265160.1 5 1104
XM_011533674.2 XP_011531976.1 7 1797
XM_011533675.2 XP_011531977.1 5 1068
XM_017006345.1 XP_016861834.1 7 1853
XM_017006346.1 XP_016861835.1 7 1796
GenBank mRNA AY262260.1 - 6 1777
AY295873.2 - 8 1974
BC101486.1 - 6 1688 8068992
BC143439.1 - 6 1653 9051947

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-96jdh:80/100.66.75.105:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline