This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DPPA3 SIRNA Assays ›
Gene Symbol:
DPPA3 DPPA3P2
Gene Name:
developmental pluripotency associated 3
developmental pluripotency associated 3 pseudogene 2
Gene Aliases:
STELLA
STELLAR
Chromosome Location:
Chr.12: 7711454 - 7717559 on Build GRCh38
Chromosome Location:
Chr.14: 36371318 - 36372369 on Build GRCh38
UniGene ID:
Hs.131358
Hs.528118
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s51494

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DPPA3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199286.3 NP_954980.1 4 498
GenBank mRNA AY317075.1 - 4 498
Gene Symbol: DPPA3P2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY230136.1 - 1 437
BC062480.1 - 1 630 30398001
BC125145.1 - 1 449 40120129

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

covalent chromatin modification
embryonic cleavage
protection of DNA demethylation of female pronucleus
negative regulation of DNA demethylation
regulation of genetic imprinting
methylated histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vvs6v:80/100.66.75.180:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0