This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C5ORF34 SIRNA Assays ›
Gene Symbol:
C5orf34
Gene Name:
chromosome 5 open reading frame 34
Gene Aliases:
-
Chromosome Location:
Chr.5: 43486699 - 43515433 on Build GRCh38
UniGene ID:
Hs.661641
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s51707

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C5orf34
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198566.3 NP_940968.1 5, 6 1425
XM_011514038.2 XP_011512340.1 5, 6 1334
XM_017009445.1 XP_016864934.1 4, 5 1456
XM_017009446.1 XP_016864935.1 3, 4 1366
GenBank mRNA AK056925.1 - 5, 6 1340
BC036867.1 - 5, 6 1288 5198039

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0