This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VGLL3 SIRNA Assays ›
Gene Symbol:
VGLL3
Gene Name:
vestigial like family member 3
Gene Aliases:
VGL-3, VGL3
Chromosome Location:
Chr.3: 86937969 - 86991293 on Build GRCh38
UniGene ID:
Hs.606507
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s52380

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: VGLL3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320493.1 NP_001307422.1 3 1246
NM_001320494.1 NP_001307423.1 3 1087
NM_016206.3 NP_057290.2 3 1246
XM_006713138.3 XP_006713201.1 3 1413
GenBank mRNA BC094780.1 - 3 1242 30528902
BX369995.2 - 2 587
CN287582.1 - 1 215
CR936700.1 - 3 882

Back To Top

More Information


Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s)

transcription, DNA-templated
regulation of transcription, DNA-templated

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-bxnfn:80/100.66.79.240:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0