This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM150A SIRNA Assays ›
Gene Symbol:
FAM150A
Gene Name:
family with sequence similarity 150 member A
Gene Aliases:
UNQ9433
Chromosome Location:
Chr.8: 52534037 - 52565461 on Build GRCh38
UniGene ID:
Hs.527111
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s52524

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM150A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207413.3 NP_997296.1 2 412
GenBank mRNA AY358213.1 - 2 415
BC130641.1 - 2 359 40146672
BC130643.1 - 2 359 40146674
BC144534.1 - 2 359 9053061

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of neuron projection development
positive regulation of protein tyrosine kinase activity
positive regulation of ERK1 and ERK2 cascade
positive regulation of ERK5 cascade
protein binding
receptor signaling protein tyrosine kinase activator activity
receptor tyrosine kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline