This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NAGS SIRNA Assays ›
Gene Symbol:
NAGS
Gene Name:
N-acetylglutamate synthase
Gene Aliases:
AGAS, ARGA
Chromosome Location:
Chr.17: 44004546 - 44009068 on Build GRCh38
UniGene ID:
Hs.8876
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s528788

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NAGS
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153006.2 NP_694551.1 7 2043
XM_011524438.1 XP_011522740.1 6 1978
XM_011524439.2 XP_011522741.1 7 1933
GenBank mRNA AK074407.1 - 7 1973
AK091938.1 - 4 1931

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

urea cycle
arginine biosynthetic process
glutamate metabolic process
acetylglutamate kinase activity
acetyl-CoA:L-glutamate N-acetyltransferase activity
arginine binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0