This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PDCD10 SIRNA Assays ›
Gene Symbol:
PDCD10
Gene Name:
programmed cell death 10
Gene Aliases:
CCM3, TFAR15
Chromosome Location:
Chr.3: 167683891 - 167735690 on Build GRCh38
UniGene ID:
Hs.478150
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s530946

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PDCD10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007217.3 NP_009148.2 6 709
NM_145859.1 NP_665858.1 6 558
NM_145860.1 NP_665859.1 5 464
XM_005247086.4 XP_005247143.1 6 1444
XM_005247087.4 XP_005247144.1 6 1449
XM_005247088.3 XP_005247145.1 5 540
XM_006713485.3 XP_006713548.1 6 604
XM_011512368.2 XP_011510670.1 7 1517
XM_011512369.2 XP_011510671.1 7 1522
XM_017005644.1 XP_016861133.1 6 599
XM_017005645.1 XP_016861134.1 6 1440
GenBank mRNA AK291130.1 - 5 495
BC002506.1 - 6 721 3050540
BC016353.1 - 6 562 4092244
CR457107.1 - 4 311

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
positive regulation of cell proliferation
positive regulation of gene expression
negative regulation of gene expression
positive regulation of cell migration
positive regulation of stress-activated MAPK cascade
positive regulation of peptidyl-serine phosphorylation
regulation of Rho protein signal transduction
intrinsic apoptotic signaling pathway in response to hydrogen peroxide
response to hydrogen peroxide
negative regulation of apoptotic process
stress fiber assembly
positive regulation of MAP kinase activity
wound healing, spreading of cells
positive regulation of Notch signaling pathway
protein stabilization
establishment of Golgi localization
positive regulation of protein serine/threonine kinase activity
negative regulation of cell migration involved in sprouting angiogenesis
Golgi reassembly
negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
protein binding
protein kinase binding
protein homodimerization activity
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline