This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ANKS6 SIRNA Assays ›
Gene Symbol:
ANKS6
Gene Name:
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases:
ANKRD14, NPHP16, PKDR1, SAMD6
Chromosome Location:
Chr.9: 98732009 - 98796542 on Build GRCh38
UniGene ID:
Hs.406890
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s532139

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ANKS6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173551.4 NP_775822.3 14 2541
XM_005251793.4 XP_005251850.1 14 2531
XM_005251794.4 XP_005251851.1 14 2528
XM_006716998.3 XP_006717061.1 14 2531
XM_006716999.2 XP_006717062.1 14 3616
XM_011518357.2 XP_011516659.1 14 4613
XM_017014445.1 XP_016869934.1 14 2528
XM_017014446.1 XP_016869935.1 13 4492
XM_017014447.1 XP_016869936.1 10 1325
GenBank mRNA AK094247.1 - 7 898
AK126614.1 - 13 1630
AK127702.1 - 13 2128
BC064367.1 - 11 1528 5562895
CR749467.1 - 14 2212
CR749472.1 - 12 1892

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline