This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EFR3B SIRNA Assays ›
Gene Symbol:
EFR3B
Gene Name:
EFR3 homolog B
Gene Aliases:
KIAA0953
Chromosome Location:
Chr.2: 25042104 - 25159137 on Build GRCh38
UniGene ID:
Hs.4892
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s533105

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: EFR3B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319099.1 NP_001306028.1 3 344
NM_014971.1 NP_055786.1 3 356
XM_011532701.2 XP_011531003.1 3 356
GenBank mRNA AB023170.1 - 3 330
AK095190.1 - 3 344
DA241650.1 - 3 356

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phosphatidylinositol phosphorylation
establishment of protein localization to plasma membrane
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jp5xp:80/100.66.72.210:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0