This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CLEC7A SIRNA Assays ›
Gene Symbol:
CLEC7A
Gene Name:
C-type lectin domain family 7 member A
Gene Aliases:
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2
Chromosome Location:
Chr.12: 10116777 - 10130269 on Build GRCh38
UniGene ID:
Hs.143929
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s535051

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CLEC7A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022570.4 NP_072092.2 3 469
NM_197947.2 NP_922938.1 4 607
NM_197948.2 NP_922939.1 4 607
NM_197949.2 NP_922940.1 3 469
NM_197950.2 NP_922941.1 2 370
NR_125336.1 4 607
XM_006719135.3 XP_006719198.1 4 602
XM_017019822.1 XP_016875311.1 4 601
XM_017019823.1 XP_016875312.1 3 366
GenBank mRNA AF313468.1 - 5 471
AF313469.1 - 3 332
AF400595.1 - 4 420
AF400596.1 - 3 282
AF400597.1 - 4 420
AF400598.1 - 3 282
AF400599.1 - 2 183
AF400601.1 - 4 420
AF400602.1 - 5 470
AJ312372.1 - 3 437
AJ312373.1 - 4 575
AK294111.1 - 4 599
AK297028.1 - 3 353
AK298679.1 - 4 523
AK298724.1 - 2 286
AK313247.1 - 3 400
AY009090.2 - 3 353
AY026769.2 - 4 420
AY026770.2 - 3 282
AY026771.1 - 3 282
AY359002.1 - 3 441
BC071746.1 - 3 383 4690475
CR749536.1 - 3 1589

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

pattern recognition receptor signaling pathway
stimulatory C-type lectin receptor signaling pathway
leukocyte activation involved in immune response
phagocytosis, recognition
inflammatory response
cell recognition
carbohydrate mediated signaling
T cell activation
defense response to protozoan
innate immune response
cellular response to molecule of fungal origin
protein binding
signaling pattern recognition receptor activity
carbohydrate binding
MHC protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-lqf76:80/100.66.77.18:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline