This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RGS7BP SIRNA Assays ›
Gene Symbol:
RGS7BP
Gene Name:
regulator of G-protein signaling 7 binding protein
Gene Aliases:
R7BP
Chromosome Location:
Chr.5: 64504565 - 64614661 on Build GRCh38
UniGene ID:
Hs.657133
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s53505

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGS7BP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029875.2 NP_001025046.1 4 1276
NM_001271890.1 NP_001258819.1 2 390
NM_001271891.1 NP_001258820.1 2 390
NR_073511.1 2 390
XM_005248502.3 XP_005248559.1 4 2658
GenBank mRNA AK294808.1 - 3 381
AK296443.1 - 4 924
AK296494.1 - 2 358
BC140707.1 - 4 622 9021575
BX640900.1 - 4 514
DA417118.1 - 2 358
DC342795.1 - 2 343

Back To Top

More Information


Gene Ontology Categories:

Function(s)

G-protein coupled receptor signaling pathway
negative regulation of signal transduction

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-grsfz:80/100.66.79.145:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0