This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IFNGR2 SIRNA Assays ›
Gene Symbol:
IFNGR2
Gene Name:
interferon gamma receptor 2 (interferon gamma transducer 1)
Gene Aliases:
AF-1, IFGR2, IFNGT1, IMD28
Chromosome Location:
Chr.21: 33402896 - 33437521 on Build GRCh38
UniGene ID:
Hs.634632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s535280

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IFNGR2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005534.3 NP_005525.2 6 1510
XM_005260969.2 XP_005261026.1 7 1026
XM_011529553.1 XP_011527855.1 7 952
XM_011529554.2 XP_011527856.1 6 1386
GenBank mRNA AK292246.1 - 6 958
AK314005.1 - 6 971
BC003624.2 - 6 978 2967074
U05875.1 - 6 1510
U05877.1 - 6 975

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
response to virus
interferon-gamma-mediated signaling pathway
regulation of interferon-gamma-mediated signaling pathway
interferon-gamma receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-49z85:80/100.66.76.252:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0