siRNA Details
| Gene Symbol: ZMYND12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146192.1 | NP_001139664.1 | 4 | 770 | ||
| NM_032257.4 | NP_115633.3 | 5 | 942 | |||
| GenBank mRNA | AK057384.1 | - | 5 | 942 | ||
| AK301839.1 | - | 4 | 756 | |||
| AL136858.1 | - | 5 | 749 | |||
| BC024186.2 | - | 5 | 931 | 4826255 | ||
| BQ441452.1 | - | 2 | 231 | |||

ZMYND12
Genomic Map