siRNA Details
| Gene Symbol: DMPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081563.2 | NP_001075032.1 | 1 | 268 | ||
| NM_001288764.1 | NP_001275693.1 | 2 | 411 | |||
| GenBank mRNA | AB209363.1 | - | 2 | 742 | ||
| AK304749.1 | - | 1 | 171 | |||
| BC026328.1 | - | 2 | 406 | 4792104 | ||
| BG706056.1 | - | 2 | 411 | |||
| M87312.1 | - | 1 | 268 | |||

DMPK
Genomic Map