siRNA Details
| Gene Symbol: WDR38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045476.2 | NP_001038941.1 | 5 | 573 | ||
| NM_001276374.1 | NP_001263303.1 | 5 | 573 | |||
| NM_001276375.1 | NP_001263304.1 | 5 | 540 | |||
| NM_001276376.1 | NP_001263305.1 | 4 | 520 | |||
| GenBank mRNA | BC127949.1 | - | 5 | 537 | 40132852 | |
| BC150645.1 | - | 5 | 573 | 9057015 | ||
| BC171828.1 | - | 5 | 573 | 9054482 | ||
| BC171830.1 | - | 5 | 540 | 9054484 | ||
| BC171837.1 | - | 4 | 520 | |||

WDR38
Genomic Map