siRNA Details
| Gene Symbol: RGS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039152.3 | NP_001034241.1 | 4, 5 | 426 | ||
| GenBank mRNA | AY643711.1 | - | 4, 5 | 427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGS21| Gene Symbol: RGS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039152.3 | NP_001034241.1 | 4, 5 | 426 | ||
| GenBank mRNA | AY643711.1 | - | 4, 5 | 427 | ||