This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CTXN3 SIRNA Assays ›
Gene Symbol:
CTXN3
Gene Name:
cortexin 3
Gene Aliases:
KABE
Chromosome Location:
Chr.5: 127649021 - 127658630 on Build GRCh38
UniGene ID:
Hs.66194
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s54778

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CTXN3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048252.2 NP_001041717.1 3 731
NM_001127385.1 NP_001120857.1 3 540
GenBank mRNA AB219764.1 - 3 708
AB219832.1 - 3 506
BC071790.1 - 3 504
BC147012.1 - 1 183 9056940

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline