siRNA Details
| Gene Symbol: MGAT4EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038135.2 | 3 | 1006 | |||
| GenBank mRNA | BC032956.1 | - | 3 | 1003 | 4827085 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGAT4EP| Gene Symbol: MGAT4EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038135.2 | 3 | 1006 | |||
| GenBank mRNA | BC032956.1 | - | 3 | 1003 | 4827085 | |