This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SMIM23 SIRNA Assays ›
Gene Symbol:
SMIM23
Gene Name:
small integral membrane protein 23
Gene Aliases:
C5orf50
Chromosome Location:
Chr.5: 171772637 - 171791136 on Build GRCh38
UniGene ID:
Hs.591740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s55889

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SMIM23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289970.1 NP_001276899.1 4 271
XM_011534622.1 XP_011532924.1 4 354
XM_011534623.2 XP_011532925.1 4 465
XM_011534624.2 XP_011532926.1 5 353
GenBank mRNA EG327678.1 - 4 216

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline