siRNA Details
| Gene Symbol: SMCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145010.1 | NP_001138482.1 | 3 | 373 | ||
| XM_011520634.2 | XP_011518936.1 | 3 | 551 | |||
| XM_011520636.2 | XP_011518938.1 | 3 | 551 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMCO2| Gene Symbol: SMCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145010.1 | NP_001138482.1 | 3 | 373 | ||
| XM_011520634.2 | XP_011518936.1 | 3 | 551 | |||
| XM_011520636.2 | XP_011518938.1 | 3 | 551 | |||