siRNA Details
| Gene Symbol: SMCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145010.1 | NP_001138482.1 | 2 | 248 | ||
| XM_011520634.2 | XP_011518936.1 | 2 | 426 | |||
| XM_011520635.2 | XP_011518937.1 | 1 | 25 | |||
| XM_011520636.2 | XP_011518938.1 | 2 | 426 | |||

SMCO2
Genomic Map