This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GCDH SIRNA Assays ›
Gene Symbol:
GCDH
Gene Name:
glutaryl-CoA dehydrogenase
Gene Aliases:
ACAD5, GCD
Chromosome Location:
Chr.19: 12891129 - 12915345 on Build GRCh38
UniGene ID:
Hs.532699
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s5637
Validated

Purification
Size
Availability Inventoried
Catalog # 4390824
Price 556.00
Your Price
Online offer: 529.65
529.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GCDH
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000159.3 NP_000150.1 10 1141
NM_013976.3 NP_039663.1 10 1141
NR_102316.1 10 1196
NR_102317.1 9 1414
XM_006722721.2 XP_006722784.1 10 1110
XM_011527899.2 XP_011526201.1 10 1110
XM_011527900.2 XP_011526202.1 10 1110
XM_017026580.1 XP_016882069.1 10 1110
GenBank mRNA AK098370.1 - 10 1110
AK290407.1 - 10 1110
AK296446.1 - 10 1159
AK298772.1 - 8 1011
AK300841.1 - 9 1363
BC002579.2 - 10 1094 3142978
BT006706.1 - 9 1033
U69141.1 - 9 1069

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00240811_m1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

lysine catabolic process
tryptophan metabolic process
fatty acid beta-oxidation using acyl-CoA dehydrogenase
fatty-acyl-CoA biosynthetic process
lipid homeostasis
fatty-acyl-CoA binding
glutaryl-CoA dehydrogenase activity
electron carrier activity
flavin adenine dinucleotide binding
oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-474fd:80/100.66.77.40:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0