This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NAV3 SIRNA Assays ›
Gene Symbol:
NAV3
Gene Name:
neuron navigator 3
Gene Aliases:
POMFIL1, STEERIN3, unc53H3
Chromosome Location:
Chr.12: 77571862 - 78213012 on Build GRCh38
UniGene ID:
Hs.655301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s57457

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NAV3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024383.1 NP_001019554.1 29 5611
NM_014903.5 NP_055718.4 28 5545
XM_005269215.3 XP_005269272.1 27 9525
XM_011538940.2 XP_011537242.1 27 9505
XM_011538941.2 XP_011537243.1 26 9483
XM_011538944.2 XP_011537246.1 26 9013
XM_017020164.1 XP_016875653.1 28 9593
XM_017020165.1 XP_016875654.1 27 9551
XM_017020166.1 XP_016875655.1 29 5600
XM_017020167.1 XP_016875656.1 28 5531
XM_017020168.1 XP_016875657.1 28 9124
XM_017020169.1 XP_016875658.1 27 9081
XM_017020170.1 XP_016875659.1 25 8991
XM_017020171.1 XP_016875660.1 21 8907
XM_017020172.1 XP_016875661.1 18 4416
XM_017020173.1 XP_016875662.1 20 8416
XM_017020174.1 XP_016875663.1 19 8374
XM_017020175.1 XP_016875664.1 19 4229
GenBank mRNA AB023155.2 - 13 2055
AF397731.1 - 27 5129
AJ488201.1 - 29 5483
AK298525.1 - 11 1238

Back To Top

More Information


Gene Ontology Categories:

Process(es)

ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-ggjf2:80/100.66.78.49:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0