This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GOLGA2P10 SIRNA Assays ›
Gene Symbol:
GOLGA2P10
Gene Name:
golgin A2 pseudogene 10
Gene Aliases:
-
Chromosome Location:
Chr.15: 82471477 - 82513999 on Build GRCh38
UniGene ID:
Hs.454640
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s57887

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GOLGA2P10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026811.3 4, 5 702
NR_033936.3 3, 4 613
NR_103496.2 3, 4 563
GenBank mRNA AK309732.1 - 3, 4 564
AK310769.1 - 3, 4 516
AK311170.1 - 3, 4 523
BC007811.2 - 4, 5 626
BC065260.1 - 4, 5 619
BC072390.1 - 3, 4 566

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline