This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF705D SIRNA Assays ›
Gene Symbol:
ZNF705D ZNF705A ZNF705E
Gene Name:
zinc finger protein 705D
zinc finger protein 705A
zinc finger protein 705E
Gene Aliases:
ZNF705C
-
-
Chromosome Location:
Chr.8: 12089338 - 12115516 on Build GRCh38
Chromosome Location:
Chr.12: 8172554 - 8180046 on Build GRCh38
Chromosome Location:
Chr.11: 71814045 - 71821547 on Build GRCh38
UniGene ID:
Hs.652228
Hs.438536
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s59194

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF705D
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039615.3 NP_001034704.2 7 1753
XM_011543832.2 XP_011542134.1 5 1535
GenBank mRNA BC110823.1 - 7 1753 6164596
Gene Symbol: ZNF705A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004328.2 NP_001004328.1 5 1403
Gene Symbol: ZNF705E
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278713.1 NP_001265642.1 5 1402

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bbcf2:80/100.66.78.33:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline