This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ANK3 SIRNA Assays ›
Gene Symbol:
ANK3
Gene Name:
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases:
ANKYRIN-G, MRT37
Chromosome Location:
Chr.10: 60026298 - 60733526 on Build GRCh38
UniGene ID:
Hs.499725
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s59467

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 374.65
374.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ANK3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204403.1 NP_001191332.1 1 111
GenBank mRNA BP245303.1 - 1 111
BX648574.1 - 1 76

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
ER to Golgi vesicle-mediated transport
plasma membrane organization
cytoskeletal anchoring at plasma membrane
signal transduction
axonogenesis
axon guidance
neuromuscular junction development
positive regulation of gene expression
positive regulation of cell communication by electrical coupling
positive regulation of sodium ion transport
magnesium ion homeostasis
neuronal action potential
positive regulation of homotypic cell-cell adhesion
Golgi to plasma membrane protein transport
regulation of potassium ion transport
establishment of protein localization
positive regulation of membrane potential
cellular response to magnesium ion
membrane assembly
protein localization to plasma membrane
maintenance of protein location in plasma membrane
protein targeting to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of membrane depolarization during cardiac muscle cell action potential
negative regulation of delayed rectifier potassium channel activity
positive regulation of sodium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
cytoskeletal adaptor activity
spectrin binding
protein binding, bridging
ion channel binding
cadherin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0