siRNA Details
| Gene Symbol: ZNF562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130031.1 | NP_001123503.1 | 1 | 44 | ||
| NM_001130032.1 | NP_001123504.1 | 1 | 44 | |||
| NM_001300885.1 | NP_001287814.1 | 1 | 44 | |||
| NM_017656.3 | NP_060126.2 | 1 | 44 | |||
| XM_005259941.3 | XP_005259998.1 | 1 | 28 | |||
| XM_005259944.3 | XP_005260001.1 | 1 | 236 | |||
| XM_005259951.3 | XP_005260008.1 | 1 | 236 | |||
| XM_005259952.3 | XP_005260009.1 | 1 | 235 | |||
| XM_011528100.2 | XP_011526402.1 | 1 | 234 | |||
| XM_011528102.2 | XP_011526404.1 | 1 | 237 | |||
| XM_017026898.1 | XP_016882387.1 | 1 | 28 | |||
| XM_017026899.1 | XP_016882388.1 | 1 | 235 | |||
| GenBank mRNA | AK297446.1 | - | 1 | 28 | ||
| AK300115.1 | - | 1 | 24 | |||
| AK303038.1 | - | 1 | 30 | |||
| AK304370.1 | - | 1 | 44 | |||
| BP221824.1 | - | 1 | 44 | |||
| DC403290.1 | - | 1 | 30 | |||
| DC405554.1 | - | 1 | 28 | |||

ZNF562
Genomic Map