siRNA Details
| Gene Symbol: SHISA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145204.2 | NP_001138676.2 | 3 | 1274 | ||
| XM_005255539.3 | XP_005255596.2 | 3 | 1275 | |||
| XM_011522642.2 | XP_011520944.1 | 3 | 1275 | |||
| GenBank mRNA | AK299610.1 | - | 4 | 704 | ||

SHISA9
Genomic Map