This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF138 SIRNA Assays ›
Gene Symbol:
ZNF138
Gene Name:
zinc finger protein 138
Gene Aliases:
pHZ-32
Chromosome Location:
Chr.7: 64794388 - 64853800 on Build GRCh38
UniGene ID:
Hs.184080
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s61207

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF138
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160183.2 NP_001153655.1 5 2592
NM_001271637.1 NP_001258566.1 4 2459
NM_001271638.1 NP_001258567.1 4 2466
NM_001271639.1 NP_001258568.1 4 2455
NM_001271640.1 NP_001258569.1 4 2514
NM_001271649.1 NP_001258578.1 2 2250
NM_006524.3 NP_006515.2 3 2377
NR_073391.1 5 2559
GenBank mRNA AK027335.1 - 1 1132
BC141847.1 - 4 2426

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-49kgl:80/100.66.79.151:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0