siRNA Details
| Gene Symbol: Gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013463.2 | NP_038491.2 | 7 | 1274 | ||
| GenBank mRNA | AK040081.1 | - | 7 | 1206 | ||
| AK054547.1 | - | 7 | 1226 | |||
| AK145011.1 | - | 7 | 1216 | |||
| AK145148.1 | - | 1274 | ||||
| AK164231.1 | - | 7 | 1207 | |||
| AK166204.1 | - | 7 | 1207 | |||
| AK166245.1 | - | 7 | 1223 | |||
| BC009021.1 | - | 7 | 1227 | 3708950 | ||
| U34071.1 | - | 7 | 1224 | |||
| U50716.1 | - | 7 | 1212 | |||

Gla
Genomic Map