siRNA Details
| Gene Symbol: Bcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009744.3 | NP_033874.1 | 8 | 1816 | ||
| GenBank mRNA | AK036975.1 | - | 8 | 1840 | ||
| AK039228.1 | - | 8 | 1838 | |||
| AK151229.1 | - | 8 | 1841 | |||
| BC052315.1 | - | 8 | 1817 | 6309948 | ||
| D38377.1 | - | 8 | 1805 | |||
| U41465.1 | - | 7 | 1828 | |||

Bcl6
Genomic Map