This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CFLAR SIRNA Assays ›
Gene Symbol:
Cflar
Gene Name:
CASP8 and FADD-like apoptosis regulator
Gene Aliases:
2310024N18Rik, A430105C05Rik, CLARP, Cash, Casper, ENSMUSG00000072980, FLAME, FLAME-1, Flip, Gm9845, I-FLICE, MRIT, c-Flip
Chromosome Location:
Chr.1: 58711310 - 58759209 on Build GRCm38
UniGene ID:
Mm.336848
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s63908

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cflar
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289704.2 NP_001276633.1 5 815
NM_001293804.1 NP_001280733.1 5 798
NM_001293805.1 NP_001280734.1 3 291
NM_009805.4 NP_033935.2 5 1087
NM_207653.5 NP_997536.1 5 1087
GenBank mRNA AK143812.1 - 5 1080
AK152515.1 - 5 782
AK161120.1 - 3 294
BC029223.1 - 5 1056 4460554
BC085105.1 - 5 810
U97076.1 - 5 753
Y14041.1 - 4 665
Y14042.1 - 4 665

Back To Top

More Information


Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of peptidase activity
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of necroptotic process
negative regulation of necroptotic process
apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway
protease binding
death receptor binding
enzyme activator activity
peptidase activator activity
protein complex binding
death effector domain binding
protein heterodimerization activity
cysteine-type endopeptidase activity involved in apoptotic signaling pathway

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline