This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DRG2 SIRNA Assays ›
Gene Symbol:
Drg2
Gene Name:
developmentally regulated GTP binding protein 2
Gene Aliases:
AI255295, AI461605
Chromosome Location:
Chr.11: 60454617 - 60468724 on Build GRCm38
UniGene ID:
Mm.41803
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s65147

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Drg2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021354.1 NP_067329.1 3 336
GenBank mRNA AJ243590.1 - 3 336
BC082564.1 - 3 292 30551418

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline