siRNA Details
| Gene Symbol: Ezh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146689.1 | NP_001140161.1 | 11 | 1462 | ||
| NM_007971.2 | NP_031997.2 | 11 | 1474 | |||
| GenBank mRNA | AK157853.1 | - | 11 | 1475 | ||
| AK220174.1 | - | 11 | 1429 | |||
| BC003772.1 | - | 11 | 1383 | 3492110 | ||
| BC016391.1 | - | 11 | 1396 | 3586689 | ||
| BC079538.1 | - | 11 | 1413 | 6817366 | ||
| U52951.1 | - | 11 | 1464 | |||

Ezh2
Genomic Map