This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GSTM4 SIRNA Assays ›
Gene Symbol:
Gstm4
Gene Name:
glutathione S-transferase, mu 4
Gene Aliases:
1110004G14Rik, GSTM7-7, Gstb-4, Gstb4
Chromosome Location:
Chr.3: 108040408 - 108045080 on Build GRCm38
UniGene ID:
Mm.31203
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s67134

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gstm4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160411.1 NP_001153883.1 4 296
NM_026764.3 NP_081040.1 5 501
GenBank mRNA AF464943.1 - 5 331
AF501320.1 - 5 317
AK003418.1 - 4 297
BC030444.1 - 5 448 5373062
CK335113.1 - 5 486

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024304, FQ223925, BC091199, FJ179403.

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
nitrobenzene metabolic process
xenobiotic catabolic process
glutathione transferase activity
enzyme binding
protein homodimerization activity
glutathione binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0