This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ITM2B SIRNA Assays ›
Gene Symbol:
Itm2b
Gene Name:
integral membrane protein 2B
Gene Aliases:
AI256040, Bri, Bri2, Bricd2b, D14Sel6, E25BMM, imBRI2
Chromosome Location:
Chr.14: 73362231 - 73385271 on Build GRCm38
UniGene ID:
Mm.392973
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s68507

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Itm2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008410.2 NP_032436.1 4 696
GenBank mRNA AB030203.1 - 4 716
AB030204.1 - 3 512
AK002599.1 - 4 659
AK003224.1 - 4 663
AK003587.1 - 4 685
AK003928.1 - 4 684
AK005125.1 - 4 687
AK076139.1 - 4 705
AK077482.1 - 4 698
AK088270.1 - 4 683
AK150378.1 - 4 716
AK150396.1 - 4 715
AK151714.1 - 4 715
AK151889.1 - 4 716
AK152068.1 - 4 704
AK152516.1 - 4 716
AK152650.1 - 4 716
AK152731.1 - 4 687
AK152983.1 - 4 708
AK153103.1 - 4 659
AK159307.1 - 4 659
AK159340.1 - 4 684
AK159378.1 - 4 715
AK159426.1 - 4 684
AK159619.1 - 4 708
AK159628.1 - 4 684
AK159792.1 - 4 684
AK159962.1 - 4 687
AK160101.1 - 4 684
AK160725.1 - 4 715
BC004731.1 - 4 667 3583154
BC010320.1 - 4 699 3599190
BC021786.1 - 4 705 4458918
U76253.1 - 4 696

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of amyloid precursor protein biosynthetic process
extrinsic apoptotic signaling pathway in absence of ligand
beta-amyloid binding
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline