This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KCNK3 SIRNA Assays ›
Gene Symbol:
Kcnk3
Gene Name:
potassium channel, subfamily K, member 3
Gene Aliases:
TASK, Task-1, cTBAK-1
Chromosome Location:
Chr.5: 30588170 - 30625270 on Build GRCm38
UniGene ID:
Mm.439936
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s68678

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnk3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010608.2 NP_034738.1 2 879
GenBank mRNA AB008537.1 - 2 745
AB013345.1 - 2 867
AF006824.1 - 2 723
AF022821.1 - 2 726
AF065162.1 - 2 963
BC032047.1 - 2 879

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_033376, AF031384, AB048824, AB048823.

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
stabilization of membrane potential
ion transmembrane transport
regulation of ion transmembrane transport
response to drug
negative regulation of cytosolic calcium ion concentration
cellular response to hypoxia
potassium ion transmembrane transport
mitophagy in response to mitochondrial depolarization
ion channel activity
voltage-gated ion channel activity
open rectifier potassium channel activity
potassium channel activity
protein C-terminus binding
potassium ion leak channel activity
protein homodimerization activity
S100 protein binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-474fd:80/100.66.77.40:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0