This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KLK1B27 SIRNA Assays ›
Gene Symbol:
Klk1b27
Gene Name:
kallikrein 1-related peptidase b27
Gene Aliases:
Gk27, Klk21l, Klk27, mGK-27
Chromosome Location:
Chr.7: 44052290 - 44056712 on Build GRCm38
UniGene ID:
Mm.445911
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s68856

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klk1b27
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020268.3 NP_064664.1 4 624
GenBank mRNA AB039275.1 - 4 626
BC132090.1 - 4 601 40130367
BC132665.1 - 4 601 40130942

Back To Top

More Information


Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mvxzb:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline