This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HSPA6 SIRNA Assays ›
Gene Symbol:
HSPA6 HSPA7
Gene Name:
heat shock protein family A (Hsp70) member 6
heat shock protein family A (Hsp70) member 7
Gene Aliases:
HSP70B'
HSP70B
Chromosome Location:
Chr.1: 161524540 - 161526897 on Build GRCh38
Chromosome Location:
Chr.1: 161606059 - 161608551 on Build GRCh38
UniGene ID:
Hs.654614
Hs.662180
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s6983

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HSPA6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002155.4 NP_002146.2 1 1611
GenBank mRNA AK093925.1 - 1 1611
AK223362.1 - 1 1611
AK295212.1 - 2 1534
AK312756.1 - 1 1611
BC035665.1 - 1 1601 5723718
X51758.1 - 1 430
Gene Symbol: HSPA7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024151.1 1 1725

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to unfolded protein
cellular response to heat
protein refolding
cellular heat acclimation
biological_process
protein binding
ATP binding
enzyme binding
heat shock protein binding
ATPase activity, coupled
unfolded protein binding
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0