siRNA Details
| Gene Symbol: Clec4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163161.1 | NP_001156633.1 | 4 | 592 | ||
| NM_010819.4 | NP_034949.3 | 4 | 595 | |||
| GenBank mRNA | AF061272.1 | - | 4 | 527 | ||
| AK089500.1 | - | 4 | 586 | |||
| AK150926.1 | - | 4 | 498 | |||
| AK155919.1 | - | 4 | 516 | |||
| AK170692.1 | - | 4 | 456 | |||
| AK171409.1 | - | 4 | 533 | |||
| BC099471.1 | - | 4 | 505 | 30845162 | ||

Clec4d
Genomic Map