siRNA Details
| Gene Symbol: Olfr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146861.1 | NP_667072.1 | 1 | 766 | ||
| GenBank mRNA | BC116994.1 | - | 1 | 831 | 40126313 | |
| BC119365.1 | - | 1 | 831 | 8734114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr9| Gene Symbol: Olfr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146861.1 | NP_667072.1 | 1 | 766 | ||
| GenBank mRNA | BC116994.1 | - | 1 | 831 | 40126313 | |
| BC119365.1 | - | 1 | 831 | 8734114 | ||