siRNA Details
| Gene Symbol: Phex | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011077.2 | NP_035207.1 | 9 | 1484 | ||
| GenBank mRNA | AK158481.1 | - | 9 | 1511 | ||
| BC137925.1 | - | 9 | 1231 | 8860944 | ||
| BC137926.1 | - | 9 | 1231 | 8860945 | ||
| EF194891.1 | - | 9 | 1018 | |||
| U49908.1 | - | 9 | 1476 | |||
| U73910.1 | - | 9 | 1484 | |||
| U75646.1 | - | 9 | 1065 | |||

Phex
Genomic Map