siRNA Details
| Gene Symbol: IGFBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253835.1 | NP_001240764.1 | 2 | 569 | ||
| NM_001553.2 | NP_001544.1 | 2 | 569 | |||
| GenBank mRNA | AF540057.1 | - | 3 | 298 | ||
| AK303915.1 | - | 2 | 571 | |||
| AK316082.1 | - | 2 | 569 | |||
| BC017201.2 | - | 2 | 557 | 3632247 | ||
| BC066339.1 | - | 2 | 569 | 5296697 | ||
| BT006654.1 | - | 2 | 535 | |||
| L19182.1 | - | 2 | 548 | |||
| S75725.1 | - | 2 | 557 | |||

IGFBP7
Genomic Map