siRNA Details
| Gene Symbol: Rab20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011227.1 | NP_035357.1 | 2 | 232 | ||
| GenBank mRNA | AB232614.1 | - | 2 | 232 | ||
| AK145566.1 | - | 2 | 409 | |||
| AK152454.1 | - | 2 | 358 | |||
| AK163743.1 | - | 2 | 372 | |||
| AK164541.1 | - | 2 | 372 | |||
| AK217678.1 | - | 2 | 177 | |||
| BC089570.1 | - | 2 | 357 | 6747570 | ||
| X80332.1 | - | 2 | 232 | |||

Rab20
Genomic Map