This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FOXK2 SIRNA Assays ›
Gene Symbol:
FOXK2
Gene Name:
forkhead box K2
Gene Aliases:
ILF, ILF-1, ILF1
Chromosome Location:
Chr.17: 82519718 - 82604607 on Build GRCh38
UniGene ID:
Hs.591140
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s7397

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FOXK2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004514.3 NP_004505.2 3, 4 934
GenBank mRNA U58196.1 - 3, 4 945
U58197.1 - 3, 4 945
U58198.1 - 3, 4 945

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
magnesium ion binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-7l792:80/100.66.78.254:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline