This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLIT1 SIRNA Assays ›
Gene Symbol:
Slit1
Gene Name:
slit homolog 1 (Drosophila)
Gene Aliases:
Slil1, mKIAA0813
Chromosome Location:
Chr.19: 41597824 - 41747275 on Build GRCm38
UniGene ID:
Mm.39155
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s73980

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slit1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015748.3 NP_056563.2 2 671
GenBank mRNA AF144627.1 - 2 696
AK122376.1 - 2 437
BC057131.1 - 2 477 6827560
BC062091.1 - 2 477 6827560

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
axonogenesis
axon guidance
motor neuron axon guidance
olfactory bulb development
tangential migration from the subventricular zone to the olfactory bulb
telencephalon cell migration
cell differentiation
retinal ganglion cell axon guidance
dorsal/ventral axon guidance
establishment of nucleus localization
neuron projection morphogenesis
negative regulation of axon extension involved in axon guidance
axon extension involved in axon guidance
negative chemotaxis
receptor binding
calcium ion binding
heparan sulfate proteoglycan binding
Roundabout binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline