This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other STRA13 SIRNA Assays ›
Gene Symbol:
Stra13
Gene Name:
stimulated by retinoic acid 13
Gene Aliases:
-
Chromosome Location:
Chr.11: 120710941 - 120713903 on Build GRCm38
UniGene ID:
Mm.259716
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s74529

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stra13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016665.2 NP_057874.2 2 116
GenBank mRNA AK076181.1 - 2 88
AK080500.1 - 2 88
AK170126.1 - 2 88
BC052335.1 - 3 366
CA320983.1 - 1 55
CJ236047.1 - 2 117
U95003.1 - 3 180
U95004.1 - 2 81
U95005.1 - 3 354

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

resolution of meiotic recombination intermediates
DNA repair
cellular response to DNA damage stimulus
cell cycle
mitotic nuclear division
replication fork processing
cell division
kinetochore assembly
DNA binding
double-stranded DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f7kpv:80/100.66.75.105:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline