This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SKP1A SIRNA Assays ›
Gene Symbol:
Skp1a
Gene Name:
S-phase kinase-associated protein 1A
Gene Aliases:
15kDa, 2610043E24Rik, 2610206H23Rik, EMC19, OCP-II, OCP2, SKP1, Tceb1l, p19A, p19Skp1
Chromosome Location:
Chr.11: 52230522 - 52246858 on Build GRCm38
UniGene ID:
Mm.42944
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s74806

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 404.00
Your Price
Online offer: 391.65
391.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Skp1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011543.4 NP_035673.3 5 451
GenBank mRNA AF083214.1 - 5 375
AK002983.1 - 5 422
AK010628.1 - 5 437
AK012498.1 - 5 410
AK014245.1 - 5 410
AK027909.1 - 5 409
AK078330.1 - 5 437
AK088339.1 - 5 410
AK152609.1 - 5 409
AK160198.1 - 5 410
AK166673.1 - 5 410
BC002115.1 - 5 394 3490159
BU841654.1 - 6 543
Z47088.1 - 5 400

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006930, NM_170679, BI552966, BI670085, Z47087, and others.
This siRNA also targets the following transcripts in Rat: NM_001007608, XM_006246244, FQ212502, FQ212745, FQ214369, and others.

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
histone H2A monoubiquitination
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline