This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other INSRR SIRNA Assays ›
Gene Symbol:
INSRR
Gene Name:
insulin receptor related receptor
Gene Aliases:
IRR
Chromosome Location:
Chr.1: 156840873 - 156858920 on Build GRCh38
UniGene ID:
Hs.248138
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s7482

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: INSRR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014215.2 NP_055030.1 17 3394
GenBank mRNA J05046.1 - 16 3010

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

transmembrane receptor protein tyrosine kinase signaling pathway
peptidyl-tyrosine phosphorylation
male sex determination
actin cytoskeleton reorganization
protein autophosphorylation
cellular response to alkaline pH
transmembrane receptor protein tyrosine kinase activity
ATP binding
phosphatidylinositol 3-kinase binding
insulin receptor substrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline